NSI Lab Solutions Microbiological Pharmaceutical Cultures, USP <51>
Comply with guidelines set by the USP for antimicrobial effectiveness testing
  • When hydrated, activity will yield at least 1 x 108 CFU/mL
  • Stable for up to 8 hours after hydration
  • Includes sterile peptone buffer for convenience
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
  • No royalty fee
Designed to conform with USP <51> specifications, culture pellet is manufactured from USP specified pure organism strain. For your convenience, each pack includes ten culture pellets individually vacuum sealed in vials and ten 1.5 mL vials of sterile peptone buffer. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Once hydrated in the supplied peptone buffer, these samples are stable for up to 8 hours. Extra activity listed on Certificate of Analysis (CoA). Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
$602.00USD / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Organism
Availability Pricing
COMPARE
A. brasiliensis
$602.00USD / Pkg of 10
COMPARE
C. albicans
$602.00USD / Pkg of 10
COMPARE
E. coli
$602.00USD / Pkg of 10
COMPARE
P. aeruginosa
$602.00USD / Pkg of 10
COMPARE
S. aureus
$602.00USD / Pkg of 10