NSI Lab Solutions Microbiological Unit Dose™ Cultures, A to E
No dilution needed—hydrate then directly inoculate media
  • Single microorganism pellet packed in a vacuum-sealed glass vial
  • Delivers less than 100 CFU per pellet
  • Less than 10% RSD lot precision
  • No more than two passages from primary culture
  • ISO 17034 accredited
  • No royalty fee
Unit Dose™ vial contains a lyophilized single microorganism culture pellet formulated to deliver less than 100 CFU. To use, simply transfer the pellet to a sterile room temperature hydration fluid. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves in less than a minute with minimum agitation and no incubation. Directly inoculate media without serial dilution for convenience. Unit Dose™ cultures are traceable and quantitatively validated Certified Reference Materials (CRMs). Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
$305.00USD / Pkg of 10
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Organism Strain
Availability Pricing
COMPARE
A. brasiliensis NCPF 2275
$305.00USD / Pkg of 10
COMPARE
B. cereus NCTC 2599
$305.00USD / Pkg of 10
COMPARE
B. subtilis NCTC 10400
$305.00USD / Pkg of 10
COMPARE
C. albicans NCPF 3179
$305.00USD / Pkg of 10
COMPARE
E. aerogenes NCTC 10006
$305.00USD / Pkg of 10
COMPARE
E. coli NCTC 12241
$305.00USD / Pkg of 10
COMPARE
E. coli NCTC 12923
$305.00USD / Pkg of 10
COMPARE
E. coli NCTC 9001
$305.00USD / Pkg of 10
COMPARE
E. faecalis NCTC 12697
$305.00USD / Pkg of 10
COMPARE
E. faecalis NCTC 775
$305.00USD / Pkg of 10
Showing 1- 10 of 10