NSI Lab Solutions Qualitative Food Microbiology Proficiency Test Swabs
Ensure state and accreditation regulation compliance with proficiency testing
  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • Hydration fluid dissolves culture pellet in less than a minute
  • Glass-free culture swab device ensures user safety
  • ISO 17043 accredited
Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. Receive your final reports within 21 days of the study close date.
Each qualitative food microbiology proficiency test comes with five Snap-Stick™ inoculated swabs and five 4” x 4” sterile square petri dishes. To activate the microorganism, simply apply pressure to the plastic stem valve and release the hydration fluid to dissolve the microorganism pellet. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves in less than a minute with minimum agitation and no incubation. Once dissolved, remove the swab from the tube and swipe across the square petri dish base. At least 2 of 5 samples swabs will test positive for the target microorganism; acceptable evaluation requires at least 4 of 5 recorded correctly with no false negatives.
If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.
$368.00USD / Pkg of 5
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Organism
Availability Pricing
COMPARE
Qualitative Listeria spp. Environmental Swab Proficiency Test; 5/PK L. ivanovii
$368.00USD / Pkg of 5
COMPARE
Qualitative Salmonella spp. Environmental Swab Proficiency Test; 5/PK Salmonella spp.
$368.00USD / Pkg of 5
COMPARE
Qualitative L. monocytogenes Environmental Swab Proficiency Test; 5/PK L. monocytogenes
$368.00USD / Pkg of 5
COMPARE
Qualitative E. coli O157:H7 Environmental Swab Proficiency Test; 5/PK E. coli
$368.00USD / Pkg of 5