NSI Lab Solutions Qualitative Hemp Microbiology Proficiency Test Cultures
Ensure state and accreditation regulation compliance with proficiency testing
  • Two studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.
For the qualitative hemp proficiency tests, at least 2 of 5 samples will test positive for the target microorganism. Acceptable evaluation requires at least 4 of 5 recorded correctly with no false negatives. These tests can be used for cultural and/or molecular test methods. These PTs are suitable for compliance with BCC and Florida as well as other USA states' statutory requirements.
If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.
$294.00 - $330.00USD / Pkg of 5
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Organism
Availability Pricing
COMPARE
Qualitative P. aeruginosa in Edible Proficiency Test; 5/PK P. aeruginosa
$329.00USD / Pkg of 5
COMPARE
Qualitative P. aeruginosa in Hemp Proficiency Test; 5/PK P. aeruginosa
$294.00USD / Pkg of 5
COMPARE
Qualitative P. aeruginosa in Oil Proficiency Test; 5/PK P. aeruginosa
$313.00USD / Pkg of 5
COMPARE
Qualitative Aspergillus Molds in Hemp Proficiency Test; 5/PK A. flavus, A. fumigatus, A. niger, A. terreus
$294.00USD / Pkg of 5
COMPARE
Qualitative Aspergillus Molds in Edible Proficiency Test; 5/PK A. flavus, A. fumigatus, A. niger, A. terreus
$329.00USD / Pkg of 5
COMPARE
Qualitative Aspergillus Molds in Oil Proficiency Test; 5/PK A. flavus, A. fumigatus, A. niger, A. terreus
$329.00USD / Pkg of 5
COMPARE
Qualitative E. coli in Hemp Proficiency Test; 5/PK E. coli
$294.00USD / Pkg of 5
COMPARE
Qualitative E. coli O157:H7 in Hemp Proficiency Test; 5/PK E. coli
$294.00USD / Pkg of 5
COMPARE
Qualitative L. monocytogenes in Edible Proficiency Test; 5/PK L. monocytogenes
$329.00USD / Pkg of 5
COMPARE
Qualitative L. monocytogenes in Hemp Proficiency Test; 5/PK L. monocytogenes
$294.00USD / Pkg of 5
COMPARE
Qualitative Salmonella in Hemp Proficiency Test; 5/PK Salmonella
$294.00USD / Pkg of 5
COMPARE
Qualitative Salmonella in Edible Proficiency Test; 5/PK Salmonella
$329.00USD / Pkg of 5
COMPARE
Qualitative Salmonella in Oil Proficiency Test; 5/PK Salmonella
$329.00USD / Pkg of 5
COMPARE
Qualitative STEC in Hemp Proficiency Test; 5/PK Shiga toxin-producing E. coli (STEC)
$294.00USD / Pkg of 5
COMPARE
Qualitative STEC in Edible Proficiency Test; 5/PK Shiga toxin-producing E. coli (STEC)
$329.00USD / Pkg of 5
COMPARE
Qualitative STEC in Oil Proficiency Test; 5/PK Shiga toxin-producing E. coli (STEC)
$329.00USD / Pkg of 5
COMPARE
Qualitative S. aureus in Edible Proficiency Test; 5/PK S. aureus
$329.00USD / Pkg of 5
COMPARE
Qualitative S. aureus in Hemp Proficiency Test; 5/PK S. aureus
$294.00USD / Pkg of 5
COMPARE
Qualitative S. aureus in Oil Proficiency Test; 5/PK S. aureus
$330.00USD / Pkg of 5
COMPARE
Qualitative Filth/Extraneous Metals in Hemp Proficiency Test; 5/PK Filth/Extraneous metals
$308.00USD / Pkg of 5
Showing 1 - 20 of 21