NSI Lab Solutions Qualitative Hemp Microbiology PT Express™ Cultures
On-demand proficiency test (PT) for when you need results fast
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.
For the qualitative hemp proficiency tests, at least 2 of 5 samples will test positive for the target microorganism. Acceptable evaluation requires at least 4 of 5 recorded correctly with no false negatives. Your PT report is available within 48 hours once you submit your results online. These tests can be used for cultural and/or molecular test methods. These PTs are suitable for compliance with BCC and Florida as well as other USA states' statutory requirements.
Hemp PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
$381.00 - $431.00USD / Pkg of 5
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Organism
Availability Pricing
COMPARE
Qualitative P. aeruginosa in Edible PT Express™; 5/PK P. aeruginosa
$429.00USD / Pkg of 5
COMPARE
Qualitative P. aeruginosa in Hemp PT Express™; 5/PK P. aeruginosa
$381.00USD / Pkg of 5
COMPARE
Qualitative P. aeruginosa in Oil PT Express™; 5/PK P. aeruginosa
$429.00USD / Pkg of 5
COMPARE
Qualitative Aspergillus Molds in Hemp PT Express™; 5/PK A. flavus, A. fumigatus, A. niger, A. terreus
$381.00USD / Pkg of 5
COMPARE
Qualitative Aspergillus Molds in Edible PT Express™; 5/PK A. flavus, A. fumigatus, A. niger, A. terreus
$429.00USD / Pkg of 5
COMPARE
Qualitative Aspergillus Molds in Oil PT Express™; 5/PK A. flavus, A. fumigatus, A. niger, A. terreus
$431.00USD / Pkg of 5
COMPARE
Qualitative E. coli in Hemp PT Express™; 5/PK E. coli
$381.00USD / Pkg of 5
COMPARE
Qualitative E. coli O157:H7 in Hemp PT Express; 5/PK E. coli
$381.00USD / Pkg of 5
COMPARE
Qualitative L. monocytogenes in Edible PT Express™; 5/PK L. monocytogenes
$429.00USD / Pkg of 5
COMPARE
Qualitative L. monocytogenes in Oil PT Express™; 5/PK L. monocytogenes
$429.00USD / Pkg of 5
COMPARE
Qualitative L. monocytogenes in Hemp PT Express™; 5/PK L. monocytogenes
$381.00USD / Pkg of 5
COMPARE
Qualitative Salmonella in Hemp PT Express™; 5/PK Salmonella
$381.00USD / Pkg of 5
COMPARE
Qualitative Salmonella in Edible PT Express™; 5/PK Salmonella
$429.00USD / Pkg of 5
COMPARE
Qualitative Salmonella in Oil PT Express™; 5/PK Salmonella
$429.00USD / Pkg of 5
COMPARE
Qualitative STEC in Hemp PT Express™; 5/PK Shiga toxin-producing E. coli (STEC)
$381.00USD / Pkg of 5
COMPARE
Qualitative STEC in Edible PT Express™; 5/PK Shiga toxin-producing E. coli (STEC)
$429.00USD / Pkg of 5
COMPARE
Qualitative STEC in Oil PT Express™; 5/PK Shiga toxin-producing E. coli (STEC)
$429.00USD / Pkg of 5
COMPARE
Qualitative S.aureus in Edible PT Express™; 5/PK S. aureus
$429.00USD / Pkg of 5
COMPARE
Qualitative S.aureus in Hemp PT Express™; 5/PK S. aureus
$381.00USD / Pkg of 5
COMPARE
Qualitative S.aureus in Oil PT Express™; 5/PK S. aureus
$429.00USD / Pkg of 5
Showing 1 - 20 of 21