Ohaus® Explorer™ EXR Series Precision Toploading Balances
High-performance balances offer intuitive interface and outstanding accuracy
  • Achieve consistent results with built-in automatic calibration
  • Manage weighing on the intuitive 7” glass touchscreen display
  • Two touchless sensors—set to activate print, zero, tare, or calibrate
  • Digital leveling bubble ensures a smooth and straightforward setup
  • RS-232, USB, and Ethernet connections provide seamless data transfer
  • Supports HID connection to PC or keyboard without drivers
  • Save up to 1000 application sample libraries to easily manage multiple sample profiles
  • Power saving features include auto dim, auto standby, and sleep mode
  • Full metal housing offers durability
Enhance your workflow with the Explorer™ EXR precision balances featuring outstanding weighing performance, intuitive touchscreen display, and advanced communication capabilities. Automatic, user-triggered, and span calibration options ensure precise measurements; the built-in automatic calibration performs self-calibration using an internal mass. Select from 11 application modes: Basic Weighing, Parts Counting, Check Counting, Percent Weighing, Check Weighing, Dynamic Weighing, Totalization, Formulation, Differential, Density Determination, Peak Hold.
Easily manage weighing operations on the 7” touchscreen display. An easy-to-clean, chemical-resistant glass panel displays vibrant colors for easy viewing. Touchless sensors located on the terminal activate often used functions such as print, zero, tare, or calibrate, simply wave your hand over the sensors to activate. Built-in RS-232, USB (A and C), and Ethernet connections provide seamless data transfer; order an optional dongle for Bluetooth® and Wi-Fi capabilities. Save and recall up to 1000 sample library records to simplify your weighing operation. Each record may contain minimum weight, sample name, sample ID, batch ID, project name, and more.
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (g) Repeatability (mg)
Availability Pricing
Ohaus EXR223 Explorer™ EXR Series Precision Top Loading Balance, 220g x 1mg, Draftshield and Touch Screen
New Products
EW-20605-20 Mfr # EXR223
COMPARE
220 0.001 1
$3,292.94USD / Each
$3,829.00USD / Each
Ohaus EXR423 Explorer™ EXR Series Precision Top Loading Balance, 420g x 1mg, Draftshield and Touch Screen
New Products
EW-20605-21 Mfr # EXR423
COMPARE
420 0.001 1
$3,850.22USD / Each
$4,477.00USD / Each
Ohaus EXR623 Explorer™ EXR Series Precision Top Loading Balance, 620g x 1mg, Draftshield and Touch Screen
New Products
EW-20605-22 Mfr # EXR623
COMPARE
620 0.001 1
$4,704.20USD / Each
$5,470.00USD / Each
Ohaus EXR1203 Explorer™ EXR Series Precision Top Loading Balance, 1200g x 1mg, Draftshield and Touch Screen
New Products
EW-20605-23 Mfr # EXR1203
COMPARE
1200 0.001 1
$5,514.32USD / Each
$6,412.00USD / Each
Ohaus EXR2202 Explorer™ EXR Series Precision Top Loading  Balance, 2200g x 0.01g, Touch Screen
New Products
EW-20605-24 Mfr # EXR2202
COMPARE
2200 0.01 10
$3,462.36USD / Each
$4,026.00USD / Each
Ohaus EXR4202 Explorer™ EXR Series Precision Top Loading  Balance, 4200g x 0.01g, Touch Screen
New Products
EW-20605-25 Mfr # EXR4202
COMPARE
4200 0.01 10
$3,980.08USD / Each
$4,628.00USD / Each
Ohaus EXR6202 Explorer™ EXR Series Precision Top Loading  Balance, 6200g x 0.01g, Touch Screen
New Products
EW-20605-26 Mfr # EXR6202
COMPARE
6200 0.01 10
$4,516.72USD / Each
$5,252.00USD / Each
Ohaus EXR8202 Explorer™ EXR Series Precision Top Loading  Balance, 8200g x 0.01g, Touch Screen
New Products
EW-20605-27 Mfr # EXR8202
COMPARE
8200 0.01 10
$4,934.68USD / Each
$5,738.00USD / Each
Ohaus EXR12202 Explorer™ EXR Series Precision Top Loading  Balance, 12200g x 0.01g, Touch Screen
New Products
EW-20605-28 Mfr # EXR12202
COMPARE
12200 0.01 10
$5,357.80USD / Each
$6,230.00USD / Each
Ohaus EXR6201 Explorer™ EXR Series Precision Top Loading  Balance, 6200g x 0.1g, Touch Screen
New Products
EW-20605-29 Mfr # EXR6201
COMPARE
6200 0.1 100
$3,756.48USD / Each
$4,368.00USD / Each
Ohaus EXR8201 Explorer™ EXR Series Precision Top Loading  Balance, 8200g x 0.1g, Touch Screen
New Products
EW-20605-30 Mfr # EXR8201
COMPARE
8200 0.1 100
$3,933.64USD / Each
$4,574.00USD / Each
Ohaus EXR10201 Explorer™ EXR Series Precision Top Loading  Balance, 10200g x 0.1g, Touch Screen
New Products
EW-20605-31 Mfr # EXR10201
COMPARE
10200 0.1 100
$4,114.24USD / Each
$4,784.00USD / Each
Showing 1- 12 of 12