Pomona® Electronics Solar Photovoltaic (PV) Test Lead Sets
Connect to test tools or clamp meters with these photovoltaic test leads
  • Ensures safe measurements on photovoltaic (PV) modules and systems
  • Connect to instruments that accept 4 mm sheathed banana plugs
  • Nickel plated contacts
Monitor solar photovoltaic (PV) systems in solar power plants with these test lead sets. Users can connect to test tools or clamp meters with two different options.
ITEM(S) ON SALE
$59.40 - $96.30USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Pomona Electronics Solar MC4 to 4 mm Test Lead Set EW-20051-06 Mfr # PVLEAD1
COMPARE
Solar MC4 to 4 mm Test Lead Set
$59.40USD / Each
$66.00USD / Each
Pomona Electronics Solar MC4 Clamp Test Lead Set EW-20051-07 Mfr # PVLEAD3
COMPARE
Solar MC4 Clamp Test Lead Set
$96.30USD / Each
$107.00USD / Each