Reagents® Sodium Chloride, Purified

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless/white crystals
  • Density: 2.17
  • Solubility information: 360 g/L at 25 °C (68 °F)
  • pH range: 5 to 9
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$26.00 - $230.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Class Weight (g) Description
Availability Pricing
Reagents Sodium Chloride, Purified, Poly Pail, 10 kg EW-62285-67 Mfr # C2313000-10KG
COMPARE
Salts 10000 Sodium Chloride, Purified, Poly Pail, 10 kg
$118.50USD / Each
Reagents Sodium Chloride, Purified, Fiber Drum, 50 lb EW-62285-68 Mfr # C2313000-50F5
COMPARE
Salts 22680 Sodium Chloride, Purified, Fiber Drum, 50 lb
$230.00USD / Each
Reagents Sodium Chloride, Purified, Wide-Mouth Container, 500 g EW-78913-69 Mfr # C2313000-500A5
COMPARE
Salts 500 Sodium Chloride, Purified, Wide-Mouth Container, 500 g
$26.00USD / Each
Reagents Sodium Chloride, Purified, Natural Poly Container, 2.5 kg EW-78919-37 Mfr # C2313000-2.5A5
COMPARE
Salts 2500 Sodium Chloride, Purified, Natural Poly Container, 2.5 kg
$53.70USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat