Retsch Mortar Grinding Sets
  • Grinds, mixes, and homogenizes hard, brittle samples up to 9 Mohs
  • Minimizes sample-to-sample contamination with easy-to-clean mortar, pestle, and scraper
Grind virtually any sample that can be processed with a mortar and pestle with this new mortar grinder mill. Variable pressure on the pestle grinds samples between the bottom edge of the pestle and the rounded mortar wall. An adjustable scraper continuously guides material toward the grinding surface and away from the mortar walls, ensuring a homogeneously ground sample.
View and add samples through two plexiglas windows on the cover. Mill accepts particles up to 8 mm dia or from 10 to 150 mL in volume; grinds sample down to less than 35 µm. Use the ON/OFF switch for continuous operation or the built-in timer for timed operation up to 60 minutes. Longer grinding time leads to smaller final particle size. Safety features include a safety switch that shuts off the power to the grinder when the lid is open and an overload switch.
To minimize sample contamination, select a grinding set that is harder than the sample to be ground. Grinding sets are available in five different materials: porcelain, corundum, agate, chrome steel, and stainless steel (SS) — order separately.
$1,080.00 - $6,205.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Material
Availability Pricing
Retsch 02.460.0016 Porcelain Mortar for Mortar Grinder Mill EW-04181-50 Mfr # 02.460.0016
COMPARE
Porcelain
$1,785.00USD / Each
Retsch 02.461.0118 Porcelain Pestle for Mortar Grinder Mill EW-04181-51 Mfr # 02.461.0118
COMPARE
Porcelain
$1,310.00USD / Each
Retsch 02.461.0116 Sintered Corundum Pestle for Mortar Grinder Mill EW-04181-53 Mfr # 02.461.0116
COMPARE
Sintered Corundum
$1,290.00USD / Each
Retsch 02.460.0017 Sintered Corundum Mortar for Mortar Grinder Mill EW-04181-54 Mfr # 02.460.0017
COMPARE
Sintered corundum
$4,305.00USD / Each
Retsch 02.460.0098 Agate Mortar for Mortar Grinder Mill EW-04181-59 Mfr # 02.460.0098
COMPARE
Agate
$6,205.00USD / Each
Retsch 02.461.0115 Agate Pestle for Mortar Grinder Mill EW-04181-61 Mfr # 02.461.0115
COMPARE
Agate
$2,205.00USD / Each
Retsch 02.460.0018 Hardened chrome steel mortar grinding set EW-04181-62 Mfr # 02.460.0018
COMPARE
Hardened Chrome Steel
$2,855.00USD / Each
Retsch 02.461.0112 Hardened Chrome Steel Pestle Grinding Set EW-04181-63 Mfr # 02.461.0112
COMPARE
Hardened Chrome Steel
$1,080.00USD / Each
Retsch 02.461.0113 Stainless Steel Pestle for Mortar Grinder Mill EW-04181-67 Mfr # 02.461.0113
COMPARE
Stainless Steel
$1,365.00USD / Each
Showing 1- 9 of 9