Value Plastics Female Luer Lug Fitting, Tee
Easily transition to a male luer fitting with directional flexibility
  • Quick and easy assembly and disassembly without the need for tools
  • Luer fittings allow for a simple yet secure connection
  • Conforms to ISO-594-1 and ISO 594-2 standards
These Value Plastics Female Luer Tee Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The tee orientation offers directional flexibility, creating a tight seal to prevent leaks or disconnection. These adapters are available in various materials to accommodate various applications and compliance requirements.
$38.40 - $50.40USD / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Regulatory Approvals Description
Availability Pricing
Value Plastics Luer Fitting, Tee Union, White Nylon, Female Luer; 25/Pk EW-50119-97 Mfr # FTLT-1
COMPARE
Nylon Non-Phthalate Luer Fitting, Tee Union, White Nylon, Female Luer; 25/Pk
$38.40USD / Pkg of 25
Value Plastics Luer Fitting, Tee Union, Polypropylene, Female Luer Lug; 25/Pk
Always in Stock
EW-50119-98 Mfr # FTLT-6005
COMPARE
Polypropylene FDA Luer Fitting, Tee Union, Polypropylene, Female Luer Lug; 25/Pk
$50.40USD / Pkg of 25
Value Plastics Luer Fitting, Tee Union, Clear Polycarbonate, Female Luer Lug; 25/Pk EW-50119-99 Mfr # FTLT-9
COMPARE
Polycarbonate Non-Phthalate Luer Fitting, Tee Union, Clear Polycarbonate, Female Luer Lug; 25/Pk
$47.10USD / Pkg of 25