Value Plastics Luer Thread Fittings, Elbow
Easily transition to a luer fitting with directional flexibility
  • Quick and easy assembly and disassembly without the need for tools
  • Luer fittings allow for a simple yet secure connection
  • Conforms to ISO-594-1 and ISO 594-2 standards
These Value Plastics Luer Thread Elbow Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The elbow orientation offers directional flexibility, creating a tight seal to prevent leaks or disconnection. These adapters are available in various materials to accommodate various applications and compliance requirements.
$43.40 - $56.40USD / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Luer Connection Type (Port 1) Luer Connection Type (Port 2)
Availability Pricing
Value Plastics Luer Fitting, Elbow Adapter, White Nylon, Male Luer x Female Luer Thread; 25/Pk EW-50120-07 Mfr # LE87-1
COMPARE
Nylon Male Luer Female Luer Thread
$43.80USD / Pkg of 25
Value Plastics Luer Fitting, Elbow Adapter, Polypropylene, Male Luer to Female Luer Thread; 25/Pk EW-50120-08 Mfr # LE87-6005
COMPARE
Polypropylene Male Luer Female Luer Thread
$56.40USD / Pkg of 25
Value Plastics Luer Fitting, Elbow Union, Polypropylene, Female Luer Thread; 25/Pk
Always in Stock
EW-50120-47 Mfr # FTLLE-6005
COMPARE
Polypropylene Female Luer Thread Female Luer Thread
$43.40USD / Pkg of 25