Weksler® Contractor Gauges
Contractor gauges for steam, water, oil, gas, or air measurement
  • 4-1/2" corrosion resistant stainless steel case and ring, plastic window
  • Accuracy is 1% full scale
  • Balanced adjustable black pointer, adjust screw in face of instrument
  • Phosphor bronze movement
  • White dial with bold black graduations and figures
  • 1/4" NPT bottom connection only
Soft soldered phosphor bronze tube with brass socket is suitable for steam, water, air, oil, gas or any medium not corrosive to copper alloys.
ITEM(S) ON SALE
$42.08USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI)
Availability Pricing
Ashcroft Weksler® 4.5" Contractor Pressure Gauge, 0 to 60 psi EW-68427-60 Mfr # EA14B
COMPARE
±1.0% full-scale 60
$42.08USD / Each
$52.60USD / Each
Ashcroft Weksler® 4.5" Contractor Pressure Gauge, 0 to 100 psi EW-68430-35 Mfr # EA14C
COMPARE
±1.0% full-scale 100
$42.08USD / Each
$52.60USD / Each