Jenway 74-Series Spectrophotometers - How to Perform a Photometrics Measurement
How to Perform a Photometrics Measurement - Video
How to Perform a Photometrics Measurement. How to Perform a Photometrics Measurement with Jenway 74-Series Visible and UV/Visible Scanning Spectrophotometers
Date: 5/30/2018
Visible and UV/Visible Scanning Spectrophotometers with CPLive™ Connectivity
The Steps
  1. Touch "Photometrics"
  2. Touch the wavelength
  3. Use the keypad to enter the required wavelength
  4. Place blank sample into the sample chamber
  5. Touch "Blank" to peform a blank measurement
The 74 Series spectrophotometers are covered with a 3 year warranty
Jenway 74 Series Visible and UV/Visible Scanning Spectrophotometer with CPLive™ Connectivity