Cole-Parmer Hydrochloric Acid, 1.00N, 500 mL

Representative image only.

Cole-Parmer Hydrochloric Acid, 1.00N, 500 mL

Cole-Parmer | Item: EW-88400-39

Reagents and solutions to meet your lab and research needs

$34.60USD / Each

$34.60USD / Each

Buy It Together

Current Item

Cole-Parmer Hydrochloric Acid, 1.00N, 500 mL
Cole-Parmer Hydrochloric Acid, 1.00N, 500 mL
8840039

$34.60USD / Each
+ =
Total Price 157.35 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Normality1
  • Purity GradeComponents all ACS
  • Concentration0.0355
  • Empirical FormulaHCl
  • ClassTitrants
  • FormLiquid
  • SynonymsMuriatic acid
  • Volume (mL)500
  • Formula Weight36.46
  • PackagingPoly Bottle
  • DOT InformationHydrochloric Acid Solution; Class 8, Packing Group III
  • Hazard SymbolCorrosive
  • UN NumberUN1789
  • CAS Number7647-01-0
  • Shelf Life (months)24
  • DescriptionHydrochloric Acid, 1.00N, 500 mL
Reviews
Q&A