NSI Lab Solutions FM-714-20 Rapid Mold CRM for 3M™ Petriflm™ Plate; 20/PK
Representative image only.
NSI Lab Solutions FM-714-20 Rapid Mold CRM for 3M™ Petriflm™ Plate; 20/PK
NSI Lab Solutions | Item: EW-05002-97
Use with 3M™ Petrifilm™ plates for increased efficiency and reliable results
$237.00USD / Pkg of 20
$237.00USD / Pkg of 20
Product Description
Key Features
  • Hydration fluid dissolves culture pellet in less than a minute
  • No more than two passages from primary culture
  • ISO 17034 accredited
  • No royalty fee
  • Compatible with 3M™ Petrifilm™ Rapid Yeast & Mold count plates
  • Pack of 20
More About this Item
Save time without sacrificing accuracy by using these microbiological reference materials with 3M™ Petrifilm™ plates. Each dehydrated pellet packaged in a vacuum-sealed glass vial contains microorganisms that can be used as controls to validate test methods. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. For the Certificate of Analysis (CoA) to be applicable, the pellet must be rehydrated and used in its entirety. Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
3M™ and Petrifilm™ are trademarks of 3M.
Specifications
  • OrganismMultiple
  • Packaging20/PK
  • Product TypeFood and Agriculture Cultures
  • Shelf Life Months (months)18
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • DescriptionRapid Mold CRM for 3M™ Petriflm™ Plate; 20/PK