NSI Lab Solutions Microbiological Food and Agriculture Cultures for Petrifilm™ Plates
Use with 3M™ Petrifilm™ plates for increased efficiency and reliable results
  • Hydration fluid dissolves culture pellet in less than a minute
  • No more than two passages from primary culture
  • ISO 17034 accredited
  • No royalty fee
Save time without sacrificing accuracy by using these microbiological reference materials with 3M™ Petrifilm™ plates. Each dehydrated pellet packaged in a vacuum-sealed glass vial contains microorganisms that can be used as controls to validate test methods. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. For the Certificate of Analysis (CoA) to be applicable, the pellet must be rehydrated and used in its entirety. Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
$189.00 - $237.00USD / Pkg of 10
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Description Packaging
Availability Pricing
COMPARE
Aerobic Lactic Acid CRM for 3M™ Petriflm™ Plate; 10/PK 10/PK
$196.00USD / Pkg of 10
COMPARE
Aerobic Lactic Acid CRM for 3M™ Petriflm™ Plate; 20/PK 20/PK
$237.00USD / Pkg of 20
COMPARE
Aerobic Count CRM for 3M™ Petriflm™ Plate; 10/PK 10/PK
$196.00USD / Pkg of 10
COMPARE
Aerobic Count CRM for 3M™ Petriflm™ Plate; 20/PK 20/PK
$237.00USD / Pkg of 10
COMPARE
Coliform Count CRM for 3M™ Petriflm™ Plate; 10/PK 10/PK
$196.00USD / Pkg of 10
COMPARE
Coliform Count CRM for 3M™ Petriflm™ Plate; 20/PK 20/PK
$237.00USD / Pkg of 20
COMPARE
E.coli/Coliform Count CRM for 3M™ Petriflm™ Plate; 10/PK 10/PK
$196.00USD / Pkg of 10
COMPARE
E.coli/Coliform Count CRM for 3M™ Petriflm™ Plate; 20/PK 20/PK
$237.00USD / Pkg of 20
COMPARE
Rapid Mold CRM for 3M™ Petriflm™ Plate; 10/PK 10/PK
$196.00USD / Pkg of 10
COMPARE
Rapid Mold CRM for 3M™ Petriflm™ Plate; 20/PK 20/PK
$237.00USD / Pkg of 20
COMPARE
Rapid Yeast CRM for 3M™ Petriflm™ Plate; 10/PK 10/PK
$196.00USD / Pkg of 10
COMPARE
Rapid Yeast CRM for 3M™ Petriflm™ Plate; 20/PK 20/PK
$189.00USD / Pkg of 20
COMPARE
Total Enterobacteriaceae CRM for 3M™ Petriflm™ Plate; 10/PK 10/PK
$196.00USD / Pkg of 10
COMPARE
Total Enterobacteriaceae CRM for 3M™ Petriflm™ Plate; 20/PK 20/PK
$237.00USD / Pkg of 20
COMPARE
Yeast & Mold CRM for 3M™ Petriflm™ Plate; 10/PK 10/PK
$196.00USD / Pkg of 10
COMPARE
Yeast & Mold CRM for 3M™ Petriflm™ Plate; 20/PK 20/PK
$237.00USD / Pkg of 20
COMPARE
Staph Express CRM for 3M™ Petriflm™ Plate; 10/PK 10/PK
$196.00USD / Pkg of 10
COMPARE
Staph Express CRM for 3M™ Petriflm™ Plate; 20/PK 20/PK
$237.00USD / Pkg of 20
Showing 1- 18 of 18