NSI Lab Solutions QCI-144B Vanadium Drinking Water PT Express™; 2/PK
Representative image only.
NSI Lab Solutions QCI-144B Vanadium Drinking Water PT Express™; 2/PK
NSI Lab Solutions | Item: EW-05005-61
On-demand proficiency test (PT) for when you need results fast
$141.00USD / Pkg of 2
$141.00USD / Pkg of 2
Product Description
Key Features
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • Two 21-mL concentrate set for determination of Vanadium
  • Each ampule produces 2 L of sample
  • Formulated in the CA-ELAP range of 5 to 50 µg/L
More About this Item
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.
These environmental chemistry drinking water proficiency tests are designed for identification of target chemicals in drinking water.
Environmental Chemistry Drinking Water PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
Specifications
  • Element/IonVanadium
  • FormLiquid
  • Volume (mL)21
  • Packaging2/PK
  • Storage SymbolStore between 15 to 30°C
  • Shelf Life (months)24
  • DescriptionVanadium Drinking Water PT Express™; 2/PK