NSI Lab Solutions Environmental Drinking Water Inorganics PT Express™

On-demand proficiency test (PT) for when you need results fast
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry drinking water proficiency tests are designed for identification of target chemicals in drinking water.

Environmental Chemistry Drinking Water PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
$93.00 - $219.00USD / Pkg of 2

25 variations of this product are available.

See All Product Options Below
25 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Perchlorate 500 Perchlorate - Whole Volume Drinking Water PT Express™; 2/PK
$119.25USD / Pkg of 2
COMPARE
Fluoride 250 Low Level Fluoride Drinking Water PT Express™; 2/PK
$97.30USD / Pkg of 2
COMPARE
Vanadium 21 Vanadium Drinking Water PT Express™; 2/PK
$141.00USD / Pkg of 2
COMPARE
pH 250 pH Drinking Water PT Express™; 2/PK
$97.30USD / Pkg of 2
COMPARE
Color 100 Color Drinking Water PT Express™; 2/PK
$145.00USD / Pkg of 2
COMPARE
Silica 21 Silica Drinking Water PT Express™; 2/PK
$109.00USD / Pkg of 2
COMPARE
Total Cyanide 21 Cyanide Drinking Water PT Express™; 2/PK
$109.00USD / Pkg of 2
COMPARE
Mercury 21 Mercury Drinking Water PT Express™; 2/PK
$99.00USD / Pkg of 2
COMPARE
Nitrate 21 Nitrate Drinking Water PT Express™; 2/PK
$93.00USD / Pkg of 2
COMPARE
Nitrite 21 Nitrite Drinking Water PT Express™; 2/PK
$105.00USD / Pkg of 2
COMPARE
TOC and DOC 21 TOC/DOC Drinking Water PT Express™; 2/PK
$120.00USD / Pkg of 2
COMPARE
Uranium 21 Uranium Drinking Water PT Express™; 2/PK
$155.00USD / Pkg of 2
COMPARE
Fluoride 125 Fluoride Drinking Water PT Express™; 2/PK
$119.00USD / Pkg of 2
COMPARE
Calcium, Magnesium, Sodium, Calcium Hardness, Total Hardness 250 Hardness Drinking Water PT Express™; 2/PK
$142.00USD / Pkg of 2
COMPARE
LAS as MBAs 10.5 MBAs Drinking Water PT Express™; 2/PK
$133.00USD / Pkg of 2
COMPARE
Turbidity 21 Turbidity Drinking Water PT Express™; 2/PK
$117.00USD / Pkg of 2
COMPARE
Alkalinity, Chloride, Conductivity, Fluoride, Nitrate as N, Nitrate/Nitrite to N, Potassium, Total Dissolved Solids 500 Inorganics Drinking Water PT Express™; 2/PK
$169.00USD / Pkg of 2
COMPARE
Corrosivity 500 Corrosivity Drinking Water PT Express™; 2/PK
$219.00USD / Pkg of 2
COMPARE
Perchlorate 5 Perchlorate Drinking Water PT Express™; 2/PK
$109.00USD / Pkg of 2
COMPARE
Aluminum, Antimony, Arsenic, Barium, Beryllium, Boron, Cadmium, Chromium, Copper, Iron, Lead, Manganese, Molybdenum, Nickel, Selenium, Silver, Thallium, Vanadium, Zinc 21 Trace Metals Drinking Water PT Express™; 2/PK
$143.00USD / Pkg of 2
Showing 1 - 20 of 25

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat