NSI Lab Solutions QCO-119B Supplemental Volatiles Wastewater PT Express™; 2/PK

Representative image only.

NSI Lab Solutions QCO-119B Supplemental Volatiles Wastewater PT Express™; 2/PK

NSI Lab Solutions | Item: EW-61203-91

On-demand proficiency test (PT) for when you need results fast

$237.00USD / Pkg of 2

$237.00USD / Pkg of 2
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • Each ampule produces at least 1 L of sample (except for VOCs)
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • Two 1.5-mL concentrate set in methanol for determination of Supplemental Volatiles

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

This sample will contain a subset of analytes from the following list:
1-Chlorohexane: 10 to 200 µg/L
1,1-Dichloropropene: 10 to 200 µg/L
1,1,1,2-Tetrachloroethane: 10 to 200 µg/L
1,1,2-Trichloro-1,2,2-trifluoroethane: 10 to 200 µg/L
1,2-Dibromo-3-chloropropane: 10 to 200 µg/L
1,2-Dibromoethane: 10 to 200 µg/L
1,2,3-Trichlorobenzene: 10 to 200 µg/L
1,2,3-Trichloropropane: 10 to 200 µg/L
1,2,4-Trimethylbenzene: 10 to 200 µg/L
1,3-Dichloropropane: 10 to 200 µg/L
1,3,5-Trichlorobenzene: 10 to 200 µg/L
1,3,5-Trimethylbenzene: 10 to 200 µg/L
1,4-Dioxane: 10 to 1000 µg/L
2-Chlorotoluene: 10 to 200 µg/L
2,2-Dichloropropane: 10 to 200 µg/L
3,3-Dimethyl-1-butanol: 5 to 500 µg/L
4-Chlorotoluene: 10 to 200 µg/L
Allyl chloride: 10 to 200 µg/L
Bromobenzene: 10 to 200 µg/L
Bromochloromethane: 10 to 200 µg/L
Chloroprene: 10 to 200 µg/L
Cyclohexanone: 10 to 200 µg/L
cis-1,4-Dichloro-2-butene: 10 to 200 µg/L
Diethyl ether: 5 to 500 µg/L
Diisopropyl ether: 5 to 200 µg/L
Ethanol: 500 to 5000 µg/L
Ethyl methacrylate: 10 to 200 µg/L
Ethyl-tert-butyl ether: 5 to 200 µg/L
Hexachlorobutadiene: 10 to 200 µg/L
Iodomethane: 10 to 200 µg/L
Isobutyl alcohol: 10 to 1000 µg/L
Isopropylbenzene: 10 to 200 µg/L
Methacrylonitrile: 10 to 200 µg/L
Methyl methacrylate: 10 to 200 µg/L
n-Butylbenzene: 10 to 200 µg/L
n-Hexane: 10 to 200 µg/L
n-Propylbenzene: 10 to 200 µg/L
p-Isopropyltoluene: 10 to 200 µg/L
Pentachloroethane: 10 to 200 µg/L
Propionitrile: 10 to 200 µg/L
sec-Butylbenzene: 10 to 200 µg/L
t-Amyl alcohol: 5 to 500 µg/L
t-Amyl methyl ether: 5 to 500 µg/L
t-Butyl alcohol: 5 to 500 µg/L
t-Butyl formate: 50 to 500 µg/L
tert-Butylbenzene: 10 to 200 µg/L
Tetrahydrofuran: 20 to 200 µg/L
trans-1,4-Dichloro-2-butene: 10 to 200 µg/L

This sample is not listed in the TNI NPW Field of Testing.

Specifications
  • MatrixMethanol solution
  • Element/IonMultiple (see the full listing under "More About This Item")
  • FormLiquid
  • Volume (mL)1.5
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8°C
  • UN Number1230
  • Shelf Life (months)24
  • DescriptionSupplemental Volatiles Wastewater PT Express™; 2/PK
Reviews
Q&A