NSI Lab Solutions Environmental Wastewater Organics PT Express™
On-demand proficiency test (PT) for when you need results fast
  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • Each ampule produces at least 1 L of sample (except for VOCs)
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.
These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.
Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.
$107.00 - $244.00USD / Pkg of 2
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Acids Wastewater PT Express™; 2/PK
$133.00USD / Pkg of 2
COMPARE
Toxaphene 1.5 Toxaphene Wastewater PT Express™; 2/PK
$117.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Volatiles Wastewater PT Express™; 2/PK
$228.00USD / Pkg of 2
COMPARE
Dicamba, 2,4-D, 2,4,5-T, Silvex, 2,4-DB, Dalapon, Dichloroprop, Dinoseb, MCPA, MCPP, Pentachlorophenol 1.5 Herbicides Wastewater PT Express™; 2/PK
$129.00USD / Pkg of 2
COMPARE
Benzene, Ethylbenzene, Toluene, m+p-Xylene, o-Xylene, Total Xylenes, MTBE, Naphthalene 1.5 BTEX by PID Wastewater PT Express™; 2/PK
$192.00USD / Pkg of 2
COMPARE
Aroclor 1016, Aroclor 1242, Aroclor 1254, Aroclor 1260 PCBs in Oil Wastewater PT Express™; 2/PK
$181.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Nitroaromatics/Nitramines in Water Wastewater PT Express™; 2/PK
$226.00USD / Pkg of 2
COMPARE
Arochlor 1016, Arochlor 1221, Arochlor 1232, Arochlor 1242, Arochlor 1248, Arochlor 1254, Arochlor 1260, Arochlor 1268 1.5 PCB in Water Wastewater PT Express™; 2/PK
$172.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Base/Neutrals Wastewater PT Express™; 2/PK
$244.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 OP Pesticides Wastewater PT Express™; 2/PK
$174.00USD / Pkg of 2
COMPARE
Gasoline range organics 1.5 Gasoline Range Organics (GRO) Wastewater PT Express™; 2/PK
$174.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Supplemental Volatiles Wastewater PT Express™; 2/PK
$237.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Low-Level PAHs Wastewater PT Express™; 2/PK
$149.00USD / Pkg of 2
COMPARE
Chlordane 1.5 Chlordane (Total) Wastewater PT Express™; 2/PK
$117.00USD / Pkg of 2
COMPARE
1,2-Dibromoethane (EDB) 1,2-Dibromo-3-chloropropane (DBCP), 1,2,3-Trichloropropane 1.5 Low-Level Halocarbons Wastewater PT Express™; 2/PK
$107.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Organochlorine Pesticides Wastewater PT Express™; 2/PK
$244.00USD / Pkg of 2
COMPARE
Diesel range organics 1.5 Diesel Range Organics (DRO) Wastewater PT Express™; 2/PK
$174.00USD / Pkg of 2
Showing 1- 17 of 17