NSI Lab Solutions QCO-135B Low-Level PAHs Wastewater PT Express™; 2/PK

Representative image only.

NSI Lab Solutions QCO-135B Low-Level PAHs Wastewater PT Express™; 2/PK

NSI Lab Solutions | Item: EW-61204-11

On-demand proficiency test (PT) for when you need results fast

$149.00USD / Pkg of 2

$149.00USD / Pkg of 2
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • Each ampule produces at least 1 L of sample (except for VOCs)
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • Two 1.5-mL concentrate set in acetonitrile for determination of PAHs by Methods 610 or 8310

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

The sample will contain at least 80% of the analytes drawn from the following list:
1-Methylnaphthalene: 2 to 20 µg/L
2-Methylnaphthalene: 2 to 20 µg/L
Acenaphthene: 2 to 20 µg/L
Acenaphthylene: 2 to 20 µg/L
Anthracene: 0.5 to 5 µg/L
Benzo(a)anthracene: 0.5 to 5 µg/L
Benzo(b)fluoranthene: 0.5 to 5 µg/L
Benzo(k)fluoranthene: 0.5 to 5 µg/L
Benzo(g,h,i)perylene: 0.5 to 5 µg/L
Benzo(a)pyrene: 0.5 to 5 µg/L
Chrysene: 0.5 to 5 µg/L
Dibenzo(a,h)anthracene: 0.5 to 5 µg/L
Fluoranthene: 0.5 to 5 µg/L
Fluorene: 2 to 10 µg/L
Indeno(1,2,3-c,d)pyrene: 0.5 to 5 µg/L
Naphthalene: 2 to 10 µg/L
Phenanthrene: 0.5 to 5 µg/L
Pyrene: 0.5 to 5 µg/L

Specifications
  • MatrixAcetonitile
  • Element/IonMultiple (see the full listing under "More About This Item")
  • FormLiquid
  • Volume (mL)1.5
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8°C
  • UN Number1648
  • Shelf Life (months)24
  • DescriptionLow-Level PAHs Wastewater PT Express™; 2/PK
Reviews
Q&A