NSI Lab Solutions UD-11288 L. innocua NCTC 11288 Unit Dose™ Culture; 10/PK
Representative image only.
NSI Lab Solutions UD-11288 L. innocua NCTC 11288 Unit Dose™ Culture; 10/PK
NSI Lab Solutions | Item: EW-05003-32
No dilution needed—hydrate then directly inoculate media
$305.00USD / Pkg of 10
$305.00USD / Pkg of 10
Product Description
Key Features
  • Single microorganism pellet packed in a vacuum-sealed glass vial
  • Delivers less than 100 CFU per pellet
  • Less than 10% RSD lot precision
  • No more than two passages from primary culture
  • ISO 17034 accredited
  • No royalty fee
  • Equivalent to ATCC 33090
More About this Item
Unit Dose™ vial contains a lyophilized single microorganism culture pellet formulated to deliver less than 100 CFU. To use, simply transfer the pellet to a sterile room temperature hydration fluid. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves in less than a minute with minimum agitation and no incubation. Directly inoculate media without serial dilution for convenience. Unit Dose™ cultures are traceable and quantitatively validated Certified Reference Materials (CRMs). Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
ATCC is a registered trademark of American Type Culture Collection.
NCTC and NCPF reference cultures obtained from the National Collection of Type Cultures and National Collection of Pathogenic Fungi, two of the culture collections of the UK Health Security Agency.
Specifications
  • OrganismL. innocua
  • Packaging10/PK
  • Product TypeUnit Dose™ Cultures
  • Shelf Life Months (months)18
  • Shipping requirementFrozen ice pack
  • Storage requirementStore between –20 and –10°C
  • StrainNCTC 11288, ATCC 33090
  • DescriptionL. innocua NCTC 11288 Unit Dose™ Culture; 10/PK