NSI Lab Solutions Microbiological Unit Dose™ Cultures, L to S
No dilution needed—hydrate then directly inoculate media
  • Single microorganism pellet packed in a vacuum-sealed glass vial
  • Delivers less than 100 CFU per pellet
  • Less than 10% RSD lot precision
  • No more than two passages from primary culture
  • ISO 17034 accredited
  • No royalty fee
Unit Dose™ vial contains a lyophilized single microorganism culture pellet formulated to deliver less than 100 CFU. To use, simply transfer the pellet to a sterile room temperature hydration fluid. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves in less than a minute with minimum agitation and no incubation. Directly inoculate media without serial dilution for convenience. Unit Dose™ cultures are traceable and quantitatively validated Certified Reference Materials (CRMs). Store in the freezer.
Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.
$305.00USD / Pkg of 10
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Organism Strain
Availability Pricing
COMPARE
L. innocua NCTC 11288, ATCC 33090
$305.00USD / Pkg of 10
COMPARE
L. monocytogenes NCTC 7973, ATCC 35152
$305.00USD / Pkg of 10
COMPARE
P. aeruginosa NCTC 12924, ATCC 9027
$305.00USD / Pkg of 10
COMPARE
S. enterica subsp. Abaetetuba NCTC 8244
$305.00USD / Pkg of 10
COMPARE
S. aureus NCTC 10788, ATCC 6538
$305.00USD / Pkg of 10
COMPARE
S. enterica Abony NCTC 6017
$305.00USD / Pkg of 10
COMPARE
S. enterica Typhimurium NCTC 12023, ATCC 14028
$305.00USD / Pkg of 10
Showing 1- 7 of 7