Cole-Parmer® Self-Cleaning Flat pH and ORP Electrodes
Flat electrode surface prevents buildup of solids Double junctions for difficult applications Annular junction reduces reference cell fouling
  • Won’t fail even in tough industrial environments
Choose from a variety of unique electrode designs for use in tough industrial environments. All models feature BNC pH connectors to match a variety of meters and controllers. Electrodes with temperature compensation feature 100 Ω RTD temperature elements with mini-audio plug connections. Flat surface does not collect suspended solids—fluid flow across the electrode provides a cleaning action that extends electrode life and improves performance. Electrodes feature a Viton® O-ring to hold glass bulb in place.
$210.00 - $323.40USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Self-cleaning flat pH electrode, PPS housing, 10-ft cable, BNC
$210.00USD / Each
COMPARE
Self-cleaning flat pH electrode, 100-Ohm Pt RTD, PPS housing, 10-ft cable, BNC
$311.85USD / Each
COMPARE
Self-cleaning flat ORP electrode, PPS housing, 10-ft cable, BNC
$323.40USD / Each