Environmental Express® Preserved Glass Bottles
EPA-approved containers with premeasured preservative already inside
  • Borosilicate glass does not have a great pH shift and can handle a wider range of temperatures than plastic
  • Ideal for sampling and storing a wide range of chemicals or mixtures in various environments
  • Containers are lot controlled, sealed with custody tape, and include a Level 1 Certificate of Analysis from an independent laboratory
  • Included screw-top caps are composed of polypropylene (PP) and lined with PTFE
Preserved glass bottles meet EPA specifications from "Guidance for Contaminant-Free Sample Containers" for metals, cyanide, fluoride, and other water quality parameters testing.
$32.35 - $768.70USD / Case of 12
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bottle Color Capacity (mL) Special Features
Availability Pricing
COMPARE
Amber 250 0.5 mL 1:1 sulfuric acid
$34.55USD / Case of 12
COMPARE
Amber 250 1 mL 1:1 sulfuric acid
$67.95USD / Case of 24
COMPARE
Amber 500 2.5 mL sodium bisulfate
$32.35USD / Case of 12
COMPARE
Amber 500 2.5 mL sulfuric acid
$53.30USD / Case of 12
COMPARE
Amber 1000 2 mL sulfuric acid
$71.80USD / Case of 12
COMPARE
Amber 1000 50 mg sodium sulfite
$62.55USD / Case of 12
COMPARE
Amber 1000 4 mL 1:1 hydrochloric acid
$64.70USD / Case of 12
COMPARE
Amber 1000 4 mL 1:1 sulfuric acid
$65.55USD / Case of 12
COMPARE
Amber 1000 5 mL hydrochloric acid
$768.70USD / Case of 12
COMPARE
Clear 1000 6.5 mL hydrochloric acid
$56.55USD / Case of 12
COMPARE
Clear 1000 2 mL sulfuric acid
$84.30USD / Case of 12
COMPARE
Clear 1000 5 mL 1:1 Sulfuric Acid
$107.90USD / Case of 12
COMPARE
Amber 500 1 mL 1:1 sulfuric acid
$41.10USD / Case of 12
Showing 1- 13 of 13