Fluke Calibration 5611T Series - PTFE encapsulated secondary thermistor probes
5611T Series – PTFE-encapsulated secondary thermistor probes: The 5611T is flexible from one end to the other, and its PTFE coating makes it immersible in acids and other harsh chemicals. This very versatile thermistor has a 3.1 mm (1/8”) tip for measuring in tight places. Its low self-heating also makes it a good choice for measurements in air.
Available for sale in the United States and Canada only.
$1,230.00 - $1,245.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
Fluke Calibration 5611T-L Thermistor PTFE Temperature Probe EW-90448-93 Mfr # 5611-TL
COMPARE
Mini Spade Lug 212 32
$1,230.00USD / Each
Fluke Calibration 2740023 Bare Thermistor Probe, PTFE, 5 Pin Din EW-90450-21 Mfr # 5611T-D
COMPARE
5-Pin DIN 212 32
$1,230.00USD / Each
Fluke Calibration 3376880 Bare Thermistor Probe, PTFEinfo-Con EW-90450-23 Mfr # 5611T-P
COMPARE
Info-Con 212 32
$1,245.00USD / Each