ICI Quick Ship Drench Shower/Eyewash Stations
Safely washes away contaminants from body and inner eye
  • Eyewash and shower operate independently
  • Overall height is 80”
All units feature a free-standing drench shower made of orange ABS plastic and an eyewash station with stainless steel bowl, four spray heads, and “push to operate” ball valve. The shower includes a rigid pull rod and easy grip handle.
$2,800.00 - $2,975.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bowl Material Mount Description
Availability Pricing
ICI Quick Ship Drench Shower/Eyewash EW-78985-16 Mfr # CPSS909
COMPARE
Stainless steel Free standing Drench Shower/Eyewash
$2,800.00USD / Each
ICI Quick Ship ADA Drench Shower/Eyewash Station EW-78985-17 Mfr # CPSSBF909
COMPARE
Stainless steel Free standing ADA Drench Shower/Eyewash Station
$2,975.00USD / Each