ICI Quick Ship Stainless Steel Garment Racks
Open wire design promotes free flow of air
  • Easy to assemble
These 304 stainless steel garment racks are designed to provide efficient storage of cleanroom garments. The racks include electro-polished 304 stainless steel hangers, spaced at 3” (7.6 cm) intervals. Unique hanger capture mechanism provides theft deterrence.
$1,620.00 - $2,905.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
ICI Quick Ship Garment Rack, Stainless Steel, Wall Mount, 24" x 36" x 7.5" EW-78985-43 Mfr # CPGRW2436S
COMPARE
7.5 304 Stainless Steel 36
$1,620.00USD / Each
ICI Quick Ship Garment Rack, Stainless Steel, Wall Mount, 24" x 48" x 7.5" EW-78985-44 Mfr # CPGRW2448S
COMPARE
7.5 304 Stainless Steel 48
$2,035.00USD / Each
ICI Quick Ship Garment Rack, Stainless Steel, Upright, 24" x 36" x 74" EW-78985-45 Mfr # CPGRU2436S
COMPARE
74 304 Stainless Steel 36
$2,370.00USD / Each
ICI Quick Ship Garment Rack, Stainless Steel, Upright, 24" x 48" x 74" EW-78985-46 Mfr # CPGRU2448S
COMPARE
74 305 Stainless Steel 48
$2,905.00USD / Each