ICI Quick Ship Wire Shelf Super Slide

Create innovative storage
  • Nickel-chrome finish with protective coating
  • Compatible with 24” deep shelves
Create innovative storage with wire super slide and hanging totes. Compatible only with 24” deep shelves. Purchase wire super slide and hanging totes separately.
$97.00 - $212.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Width (in) Description
Availability Pricing
ICI Quick Ship Wire Super Slide; for 24" D units EW-78982-52 Mfr # CPSS2NC
COMPARE
10 1/4 20 3/8 ICI Quick Ship Wire Super Slide; for 24" D units
$212.00USD / Each
ICI Quick Ship Metro Tote, 3" H, 17 1/2" W, 22 1/2" L EW-78982-53 Mfr # CPMTB93030W
COMPARE
3 17 1/2 ICI Quick Ship Metro Totes, 3" H, 17 1/2" W, 22 1/2" L
$97.00USD / Each
ICI Quick Ship Metro Tote, 6" H, 17 1/2" W, 22 1/2" L EW-78982-54 Mfr # CPMTB93060W
COMPARE
6 17 1/2 ICI Quick Ship Metro Totes, 6" H, 17 1/2" W, 22 1/2" L
$100.00USD / Each
ICI Quick Ship Metro Tote, 8" H, 17 1/2" W, 22 1/2" L EW-78982-55 Mfr # CPMTB93080W
COMPARE
8 17 1/2 ICI Quick Ship Metro Totes, 8" H, 17 1/2" W, 22 1/2" L
$118.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat