Imada Digital Torque Gauge
FEATURES
  • Programmable high/low setpoints with LED indicators for quick GO/NO GO testing
  • 256 Data memory for recall and download
  • Accuracy: ±0.5% F.S. ±1 LSD
  • lbf-in, kgf-cm, and N-cm (selectable)
  • Outputs: RS-232, Digimatic, analog
  • 200% overload capacity
  • Ergonomic, heavy duty metal case
  • Real time and peak mode (selectable)
  • Tare Function
  • Clockwise and counterclockwise operation
  • Runs on internal NiCad batteries or AC adapter
  • Overload alarm, low battery indicator
  • Carrying case, standard attachments and AC adapter/charger included
ITEM(S) ON SALE
$1,596.25 - $1,823.75USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity
Availability Pricing
Imada HTG2-15 Digital Torque Gauge, 15 lbf-in EW-59850-27 Mfr # HTGS-15
COMPARE
15 lb in
$1,596.25USD / Each
$1,705.00USD / Each
Imada HTG2-4 Digital Torque Gauge, 70 ozf-in EW-59850-28 Mfr # HTGS-4
COMPARE
70 oz in
$1,823.75USD / Each
$1,950.00USD / Each
Imada HTG2-40 Torq Gauge 40LBF Capacity Digital EW-59850-29 Mfr # HTGS-40
COMPARE
40 lb in
$1,596.25USD / Each
$1,705.00USD / Each
Imada HTGS-85 Torque Gauge 85LBF Capacity Digital EW-59850-30 Mfr # HTGS-85
COMPARE
85 lb in
$1,711.25USD / Each
$1,830.00USD / Each