NSI Lab Solutions Environmental Wastewater Organics Proficiency Tests
Ensure state and accreditation regulation compliance with proficiency testing
  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Supplied in duplicate for convenience and value
  • Each ampule produces at least 1 L of sample (except for VOCs)
  • ISO 17043 accredited
Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.
These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.
If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.
$80.00 - $179.00USD / Pkg of 2
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Acids Wastewater Proficiency Test; 2/PK
$88.00USD / Pkg of 2
COMPARE
Aroclor 1016, Aroclor 1242, Aroclor 1254, Aroclor 1260 PCBs in Oil Wastewater Proficiency Test; 2/PK
$125.00USD / Pkg of 2
COMPARE
Toxaphene 1.5 Toxaphene Wastewater Proficiency Test; 2/PK
$80.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Volatiles Wastewater Proficiency Test; 2/PK
$160.00USD / Pkg of 2
COMPARE
Dicamba, 2,4-D, 2,4,5-T, Silvex, 2,4-DB, Dalapon, Dichloroprop, Dinoseb, MCPA, MCPP, Pentachlorophenol 1.5 Herbicides Wastewater Proficiency Test; 2/PK
$84.00USD / Pkg of 2
COMPARE
Chlordane 1.5 Chlordane (Total) Wastewater Proficiency Test; 2/PK
$80.00USD / Pkg of 2
COMPARE
Benzene, Ethylbenzene, Toluene, m+p-Xylene, o-Xylene, Total Xylenes, MTBE, Naphthalene 1.5 BTEX by PID Wastewater Proficiency Test; 2/PK
$135.00USD / Pkg of 2
COMPARE
Arochlor 1016, Arochlor 1221, Arochlor 1232, Arochlor 1242, Arochlor 1248, Arochlor 1254, Arochlor 1260, Arochlor 1268 1.5 PCB in Water Wastewater Proficiency Test; 2/PK
$135.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Base/Neutrals Wastewater Proficiency Test; 2/PK
$168.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 OP Pesticides Wastewater Proficiency Test; 2/PK
$122.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Low-Level PAHs Wastewater Proficiency Test; 2/PK
$104.00USD / Pkg of 2
COMPARE
1,2-Dibromoethane (EDB) 1,2-Dibromo-3-chloropropane (DBCP), 1,2,3-Trichloropropane 1.5 Low-Level Halocarbons Wastewater Proficiency Test; 2/PK
$84.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Supplemental Volatiles Wastewater Proficiency Test; 2/PK
$179.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Nitroaromatics/Nitramines in Water Wastewater Proficiency Test; 2/PK
$159.00USD / Pkg of 2
COMPARE
Multiple (see the full listing under "More About This Item") 1.5 Organochlorine Pesticides Wastewater Proficiency Test; 2/PK
$168.00USD / Pkg of 2
COMPARE
Diesel range organics 1.5 Diesel Range Organics (DRO) Wastewater Proficiency Test; 2/PK
$122.00USD / Pkg of 2
COMPARE
Gasoline range organics 1.5 Gasoline Range Organics (GRO) Wastewater Proficiency Test; 2/PK
$122.00USD / Pkg of 2
Showing 1- 17 of 17