NSI Lab Solutions Quantitative Food Microbiology Proficiency Test Cultures
Ensure state and accreditation regulation compliance with proficiency testing
  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.
These food microbiology proficiency tests provide the quantitative determination of target microorganisms by standardized cultural methods. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation. If applicable add meat or dairy matrix to the solution.
If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.
$189.00 - $623.00USD / Pkg of 2
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Description Packaging
Availability Pricing
COMPARE
Quantitative B. cereus with Buffer Matrix Proficiency Test 2/PK
$222.00USD / Pkg of 2
COMPARE
Quantitative Indicators with Buffer Matrix Proficiency Test; 2/PK 2/PK
$368.00USD / Pkg of 2
COMPARE
Quantitative Indicators with Buffer Matrix Proficiency Test; 10/PK 10/PK
$594.00USD / Pkg of 10
COMPARE
Quantitative Indicators with Buffer Matrix Proficiency Test; 5/PK 5/PK
$403.00USD / Pkg of 5
COMPARE
Quantitative Indicators with Dairy Matrix Proficiency Test; 2/PK 2/PK
$386.00USD / Pkg of 2
COMPARE
Quantitative Indicators with Dairy Matrix Proficiency Test; 10/PK 10/PK
$623.00USD / Pkg of 10
COMPARE
Quantitative Indicators with Dairy Matrix Proficiency Test; 5/PK 5/PK
$426.00USD / Pkg of 5
COMPARE
Quantitative Indicators with Meat Matrix Proficiency Test; 2/PK 2/PK
$386.00USD / Pkg of 2
COMPARE
Quantitative Indicators with Meat Matrix Proficiency Test; 5/PK 5/PK
$426.00USD / Pkg of 5
COMPARE
Quantitative Lactic Acid Bacteria with Buffer Matrix Proficiency Test 2/PK
$233.00USD / Each
COMPARE
Quantitative P. aerugwithosa with Buffer Matrix Proficiency Test 2/PK
$222.00USD / Each
COMPARE
Quantitative Lactic Acid Bacteria with Dairy Matrix Proficiency Test 2/PK
$233.00USD / Pkg of 2
COMPARE
Quantitative P. aerugwithosa with Dairy Matrix Proficiency Test 2/PK
$233.00USD / Pkg of 2
COMPARE
Quantitative Lactic Acid Bacteria with Meat Matrix Proficiency Test 2/PK
$189.00USD / Pkg of 2
COMPARE
Quantitative B. cereus with Dairy Matrix Proficiency Test 2/PK
$233.00USD / Pkg of 2
COMPARE
Quantitative B. cereus with Meat Matrix Proficiency Test 2/PK
$233.00USD / Pkg of 2
COMPARE
Quantitative Indicators with Meat Matrix Proficiency Test, 10/PK 10/PK
$623.00USD / Pkg of 10
COMPARE
Quantitative P. aerugwithosa with Meat Matrix Proficiency Test 2/PK
$233.00USD / Pkg of 2
Showing 1- 18 of 18