Test Products Digital Pressure Manometers
  • Measure pressure of air and natural gas
  • 655L measures pressure of non-corrosive water/liquid applications
  • Rugged extruded aluminum housing
  • Measure positive differential pressure
  • Push button zero for altitude correction
  • Seven selectable units of measure to choose from: mmHg, inHg, kPa, mBar, PSI, inH20, and mmH20
  • Backlight with auto shut off after 10 seconds
  • Auto shut off after 30 minutes (except REC mode) to save battery life
  • Easy-to-read LCD
  • Record function stores maximum and minimum readings
  • 3-year limited warranty
  • Data hold to freeze reading on LCD
    APPLICATIONS
  • Check for proper pressure in pneumatic systems
  • Check supply gas pressures
  • Check for proper pressures in clean rooms
  • Check industrial HVAC systems
  • Check operation of industrial pressure switches
$288.75 - $457.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSID) Min Pressure (PSID) Description
Availability Pricing
COMPARE
100 -100 Digital Manometer with Dual Input, 7 Selectable Units of Measure, +/-101.5 psi
$323.50USD / Each
TPI 665L Dual Input Differential Liquid Manometer, +/-100 psi EW-42500-11 Mfr # 665L
COMPARE
100 -100 Dual Input Differential Liquid Manometer, +/-100 psi
$457.00USD / Each
COMPARE
5 -5 Digital Manometer, 0/5psid Dual Input, 7 Selectable Units of Measure
$288.75USD / Each
COMPARE
30 -30 Digital Manometer, 30 psid Dual Input, 7 Selectable Units of Measure
$288.75USD / Each
TPI 645NPT Dual Input Digital Differential Manometer Kit, +/-5 psi EW-42500-25 Mfr # 645NPT
COMPARE
30 -30 Dual Input Digital Differential Manometer Kit, +/-5 psi
$323.50USD / Each