Product Type
In Stock
Industry
Max Flow Rate (CFM)
Max Flow Rate (Liters/min)
Free-Air Capacity (LPM)
Free-Air Capacity (LPH)
Free-Air Capacity (CFM)
US Origin
Vacuum Pumps
The product searched is discontinued, please browse below for replacements.
Vacuum Pumps
Reliable Vacuum Pumps for Laboratory and Industrial Applications
Enhance research accuracy and manufacturing efficiency with high-performance vacuum pumps from Cole-Parmer. Engineered for precision, our pumps support everything from portable field research to heavy industrial processing. Choose from quiet vacuum pumps ideal for sensitive laboratory environments, or robust, high-capacity vacuum pumps perfect for demanding industrial operations. Our corrosion-resistant vacuum pumps effectively manage chemical processing needs, while our compact and portable vacuum pumps offer flexibility for on-the-go requirements. Experience effortless vacuum pump integration with our comprehensive solutions, including easy-to-maintain maintenance-free vacuum pumps designed for continuous, hassle-free use. MORE
Vacuum Pumps, pressure pumps, cover a wide range of operating pressures. Laboratory/filtration vacuum pumps are for use in the lab or anywhere a low vacuum is needed; typically offer both vacuum and pressure capabilities. Use rough vacuum pumps (or roughing pumps) for laboratory and industrial applications requiring a level of vacuum less than 10–3 Torr. Use high vacuum pressure pumps when you need vacuums higher than 10–3 Torr. Also, consider the free-air capacity needed (the higher the free air capacity, the faster it will evacuate the chamber) and whether you need a lubricated (oiled) or non-lubricated (dry or oilless) pump. Lubricated pumps provide higher capacities, higher vacuum levels, and lower noise, but can contaminate the system and require more maintenance. Select a nonlubricated pump for vacuum in a clean system and low maintenance.
Cole-Parmer® Vacuum Aspiration Systems
APPLY FILTERS
filter byclear all
58 RESULTS
SORT BY
Showing 1 - 24 of 58
23>
Why Choose Cole-Parmer Vacuum Pumps?
Cole-Parmer provides precision-engineered vacuum pumps offering reliable performance, minimal maintenance, and full compliance with industry regulations.
Extensive Selection
Our portfolio includes versatile diaphragm vacuum pumps, high-capacity rotary vane vacuum pumps, and integrated vacuum systems for labs.
Proven Reliability
Designed and rigorously tested, Cole-Parmer pumps guarantee reliable operation, reduced downtime, and increased productivity.
Regulatory Compliance
All pumps adhere strictly to industry safety and compliance standards, ensuring secure and efficient operation in regulated environments.