Fluke® 1770 Series Three-Phase Power Quality Analyzers
Measure, log, and analyze power quality parameters on-the-go with one portable device
  • Automatically measure power and power quality parameters
  • Quickly identify problems while troubleshooting with live on-screen data in the field
  • Easily view V/A/Hz, power, dips, swells, and harmonics data on a large color display
  • Power directly from measurement circuit without a line cord
  • Instantly view, download, and share logged data with Fluke Energy Analyze Plus software
The Fluke® 1770 Series power quality analyzers are 2-in-1 devices that combine the troubleshooting functionality of a power quality meter with the robust analysis and logging capabilities of a standalone power quality analyzer—in a single, easy-to-use, handheld device. Features include automatic measurements, an easy-to-use interface and setup, and a simplified reporting platform. The instrument can also be powered directly from the measurement circuit, eliminating the need to find a power outlet or use a lengthy extension cord.
The unique automatic measurement capture system ensures you’re collecting the right data every time, while still giving you the flexibility to select and adjust specific parameters as needed. More than 500 power quality parameters are captured by default and the guided setup makes it easy to select the right parameters for the system you’re working on. Logged data is instantly viewable, downloadable, and shareable with Fluke Energy Analyze Plus software. The ‘PQ Meter’ function gives you immediate access to live on-screen data in the field so you can quickly identify potential problems while troubleshooting. The detailed ‘PQ Logging and Analysis’ function eliminates the complexity of performing power quality studies by guiding you through the setup process ensuring you’re capturing the right data every time.
The Fluke 1775 and 1777 analyzers incorporate advanced transient capture technology to help you clearly identify high-speed voltage transients. The 1775 analyzer has 1 MHz sampling capability to capture fast transients, while the 1777 analyzer has 20 MHz sampling capability to capture the fastest transients in high detail. NOTE: BASIC models do not come with iFlex® current clamp probes.
$8,020.00 - $14,530.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Communication Ports Max AC Current (amps) Description
Availability Pricing
Fluke 1775/FPC Three-Phase Power Quality Analyzer with 1 Yr Premium Care & Current Probes EW-20051-29 Mfr # 5335300
COMPARE
Ethernet, USB type A, USB-C, WiFi, Bluetooth 6000 1775/FPC Three-Phase Power Quality Analyzer with 1 Yr Premium Care & Current Probes
$13,000.00USD / Each
Fluke 1773 Three-Phase Power Quality Analyzer with Current Probes EW-30028-29 Mfr # 5117269
COMPARE
Ethernet, USB type A, USB-C 6000 1773 Three-Phase Power Quality Analyzer with Current Probes
$8,850.00USD / Each
Fluke 1773/BASIC Three-Phase Power Quality Analyzer without Current Probes EW-30028-30 Mfr # 5272757
COMPARE
Ethernet, USB type A, USB-C 6000 1773/BASIC Three-Phase Power Quality Analyzer without Current Probes
$8,020.00USD / Each
Fluke 1775 Three-Phase Power Quality Analyzer with Current Probes EW-30028-31 Mfr # 5117278
COMPARE
Ethernet, USB type A, USB-C, WiFi, Bluetooth 6000 1775 Three-Phase Power Quality Analyzer with Current Probes
$11,630.00USD / Each
Fluke 1775/BASIC Three-Phase Power Quality Analyzer without Current Probes EW-30028-32 Mfr # 5272784
COMPARE
Ethernet, USB type A, USB-C 6000 1775/BASIC Three-Phase Power Quality Analyzer without Current Probes
$10,800.00USD / Each
Fluke 1777 Three-Phase Power Quality Analyzer with Current Probes EW-30028-33 Mfr # 5126630
COMPARE
Ethernet, USB type A, USB-C, WiFi, Bluetooth 6000 1777 Three-Phase Power Quality Analyzer with Current Probes
$14,530.00USD / Each
Fluke 1777/BASIC Three-Phase Power Quality Analyzer without Current Probes EW-30028-34 Mfr # 5272819
COMPARE
Ethernet, USB type A, USB-C 6000 1777/BASIC Three-Phase Power Quality Analyzer without Current Probes
$13,700.00USD / Each
Showing 1- 7 of 7