Fluke® 190 Series III ScopeMeter® Test Tools
Combine rugged portability with the high performance of bench oscilloscopes
  • Rated for industrial environments CAT III 1000 V/CAT IV 600 V
  • Automatically capture, view and analyze complex waveforms
  • USB and Wi-Fi download for analyzing data with FlukeView® software
  • Large, bright color display is easy to view in nearly any environment
  • Easy to store and view historical data and transfer to a PC via USB or Wi-Fi
  • IP51 rating, dust and drip-proof
  • Frequency spectrum using FFT-analysis
The Fluke® 190 Series III ScopeMeter® is a handheld digital oscilloscope features advanced connectivity options to make data analysis, data sharing, and maintenance or troubleshooting decisions faster and easier. Included Wi-Fi adapter communicates with PCs and mobile devices on wireless LAN using the downloadable Fluke Connect app. A mini-USB-B is provided which allows for interconnection to PC for remote control and data transfer under PC-control using FlukeView-2. Choose a ScopeMeter kit for FlukeView® software, which enables further collaboration, data analysis, and archiving of critical test information.
Choose from two or four channel models with a wide range of bandwidth options. Fast sampling rates up to 5.0 GS/s, 200 ps resolution and deep memory of 10,000 samples per channel allow high-accuracy capture and display of waveform details, noise, and other disturbances. Perform timing or amplitude related measurements on three phases or three-axis control systems, or simply compare and contrast multiple test points in a circuit under test. Features like TrendPlot™ Paperless Recorder, ScopeRecord™ Mode, Connect-and-View™ Triggering and a unique 100-screen Replay function help you quickly diagnose issues to minimize repair costs and downtime.
$4,711.00 - $8,552.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bandwidth (MHz) Channels Description
Availability Pricing
Fluke 190 Series III Color ScopeMeter®, 2 Channel, 60 MHz EW-20041-70 Mfr # FLUKE-190-062-III
COMPARE
60 2 Color ScopeMeter®, 2 Channel, 60 MHz
$4,711.00USD / Each
Fluke 190 Series III Color ScopeMeter®,  2 Channel, 200 MHz EW-20041-76 Mfr # FLUKE-190-202-III
COMPARE
200 2 Color ScopeMeter®, 2 Channel, 200 MHz
$5,873.00USD / Each
Fluke 190 Series III Color ScopeMeter®,  4 Channel, 500 MHz EW-20041-82 Mfr # FLUKE-190-504-III
COMPARE
500 4 Color ScopeMeter®, 4 Channel, 500 MHz
$7,869.90USD / Each
Fluke 190 Series III Color ScopeMeter® with 1 Yr Premium Care, 4 Channel, 500 MHz EW-20051-27 Mfr # FLUKE-190-504/FPC
COMPARE
500 4 Color ScopeMeter® with 1 Yr Premium Care, 4 Channel, 500 MHz
$8,552.00USD / Each