Master® Industrial HG-Series Heat Guns
  • Flameless heat - ideal for drying specimens and beakers, or controlling heat treatment of samples and heat-shrink tubing
  • These industrial-grade heat guns feature high temperature and power ratings: up to 1200°F/649°C
Guns are powerful enough to produce an air velocity of 3400 ft./min and an air volume of 27 cfm - great whenever you need fast, flameless heat. Air blast temperature is measured at the nozzle of the heat gun. Order attachments and replacement heating element kits separately.
$255.00 - $282.40USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Air Volume (CFM) Max Temperature (° F) Power (VAC)
Availability Pricing
Master Appliance HG-201D 400°F Heat Gun, 600 Watts, 120 V, 60 Hz EW-03020-00 Mfr # HG-201D
COMPARE
27 400 120
$265.90USD / Each
Master Appliance HG-202D-02 400°F Heat Gun, 660 Watts, 220 V, 50 Hz EW-03021-00 Mfr # HG-202D-02
COMPARE
27 400 220
$282.40USD / Each
Master Appliance HG-301D 800°F Heat Gun, 1440 Watts, 120 V, 60 Hz EW-03022-00 Mfr # HG-301D
COMPARE
27 800 120
$255.00USD / Each
Master Appliance HG-302D-02 800°F Heat Gun, 1320 Watts, 220 V, 50 Hz EW-03023-00 Mfr # HG-302D-02
COMPARE
27 800 220
$261.70USD / Each
Master Appliance HG-501D 1200°F Heat Gun, 1740 Watts, 120 V, 60 Hz EW-03028-00 Mfr # HG-501D
COMPARE
27 1200 120
$275.70USD / Each
Master Appliance HG-502D-02 1200°F Heat Gun, 1980 Watts, 220 V, 50 Hz
Always in Stock
EW-03029-00 Mfr # HG-502D-02
COMPARE
27 1200 220
$265.90USD / Each