Brand

Product Type

In Stock

Normality

Industry

Organism

Strain

Chemistry

Vapor Pressure (torr)

US Origin

Standards and Solutions

The product searched is discontinued, please browse below for replacements.
Standards and Solutions
We offer a wide variety of standards and solutions to meet your chemical needs. Choose from single and multi-element ICP standards, AA standards, plasma emission standards, conductivity standards, turbidity standards, pH buffers, and normality and molarity solutions. When you need silver nitrate (solution), sodium thiosulfate, iodine and more, buy from Cole-Parmer; the one source you can trust that only carries reliable, leading brands.
Need Equipment and Supplies Now?

Need It Now?

Use the left rail availability filter to

APPLY FILTERS
filter byclear all
 
3,174 RESULTS
SORT BY
Showing 1 - 24 of 3,174
23133>